Novus Biologicals
Manufacturer Code:NBP198475
Catalog # NBP198475
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this antibody is Renalase - middle region. Peptide sequence GMKIGVPWSCRYLSSHPCICFISIDNKKRNIESSECGPSVVIQTTVPFGV. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-NAD(P)H oxidase/anomerase; C10orf59chromosome 10 open reading frame 59 EC 1.4 FLJ11218 MAO-C Monoamine oxidase-C RENALASE renalase FAD-dependent amine oxidase; MAO-C; Monoamine oxidase-C; Renalase; renalase, FAD-dependent amine oxidase
Gene Aliases: C10orf59; RENALASE; RNLS
UniProt ID: (Human) Q5VYX0
Entrez Gene ID: (Human) 55328
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.