Invitrogen
Manufacturer Code:PA579924
Catalog # PIPA579924
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence of human Regucalcin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory protein Li 41; Gluconolactonase; GNL; RC; Regucalcin; Regucalcin RC Gluconolactonase GNL Senescence marker protein 30 SMP-30 RGN SMP30; Senescence marker protein 30; SMP-30
Gene Aliases: AI265316; GNL; HEL-S-41; RC; Reguc; RGN; SMP30
UniProt ID: (Human) Q15493, (Rat) Q03336, (Mouse) Q64374
Entrez Gene ID: (Human) 9104, (Rat) 25106, (Mouse) 19733
Molecular Function:
calcium-binding protein
esterase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.