Novus Biologicals
Manufacturer Code:NBP294851100UL
Catalog # NB024211
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-299 of human RGN (NP_004674.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.1.17 Gluconolactonase GNL RCsenescence marker protein-30 regucalcin (senescence marker protein-30) Senescence marker protein 30 SMP-30 SMP30regucalcin; epididymis secretory protein Li 41; Gluconolactonase; GNL; RC; Regucalcin; Senescence marker protein 30; SMP-30
Gene Aliases: GNL; HEL-S-41; RC; RGN; SMP30
UniProt ID: (Human) Q15493
Entrez Gene ID: (Human) 9104
Molecular Function:
calcium-binding protein
esterase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.