Novus Biologicals
Manufacturer Code:NBP289832AF594
Catalog # NB109624
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 244 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human Reg4 (NP_001152824.1 Met 1-Pro 158). |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Gastrointestinal secretory protein; Gastrointestinal secretory protein GISPREG-4 Reg IV regenerating gene type IV regenerating islet-derived family member 4 regenerating islet-derived protein 4 Regenerating islet-derived protein IV REG-IV RELPREG-like protein; Reg IV; REG-4; REG-like protein; regenerating gene type IV; regenerating islet-derived family, member 4; Regenerating islet-derived protein 4; Regenerating islet-derived protein IV
Gene Aliases: GISP; REG-IV; REG4; RELP
UniProt ID: (Human) Q9BYZ8
Entrez Gene ID: (Human) 83998
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.