R&D Systems
Manufacturer Code:BAF1379
Catalog # BAF1379
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Reg4 Asp23-Pro158 Accession # Q9BYZ8 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Gastrointestinal secretory protein; Gastrointestinal secretory protein GISP GISPREG-4 Reg IV regenerating gene type IV regenerating islet-derived family member 4 regenerating islet-derived protein 4 Regenerating islet-derived protein IV REG-IV RELP RELPREG-like protein; Reg IV; REG-4; REG-like protein; regenerating gene type IV; regenerating islet-derived family, member 4; Regenerating islet-derived protein 4; Regenerating islet-derived protein IV
Gene Aliases: GISP; REG-IV; REG4; RELP
UniProt ID: (Human) Q9BYZ8
Entrez Gene ID: (Human) 83998
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.