Abnova
Manufacturer Code:MAB18728
Catalog # 89-338-653
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S345-51 |
Immunogen | Recombinant protein corresponding to amino acids 111-201 of mouse Reep1. |
Conjugate | ATTO488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AW558420; C87808; D6Ertd253e; Receptor expression-enhancing protein 1
Gene Aliases: D6Ertd253e; Reep1
UniProt ID: (Mouse) Q8BGH4
Entrez Gene ID: (Mouse) 52250
Molecular Function:
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.