Invitrogen
Manufacturer Code:PA5104090
Catalog # PIPA5104090
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RELN corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ETL7 extracellular matrix serine protease LIS2 PRO1598 Reelen reeler Reeler protein Reelin Reln Rl; extracellular matrix serine protease; Reeler protein; Reelin
Gene Aliases: ETL7; LIS2; PRO1598; Reelen; reeler; RELN; RL
UniProt ID: (Human) P78509, (Mouse) Q60841, (Rat) P58751
Entrez Gene ID: (Human) 5649, (Mouse) 19699, (Rat) 24718
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.