Invitrogen
Manufacturer Code:PA517202
Catalog # PIPA517202
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids near the amino terminus of human RecQ4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent DNA helicase Q4; ATP-dependent DNA helicase Q4 DNA helicase RecQ-like type 4 DNA helicase RecQ-like type 4; DNA helicase, RecQ-like type 4; DNA helicase, RecQ-like, type 4; RecQ helicase-like 4; RecQ protein-like 4; RecQ4; RTS
Gene Aliases: RECQ4; RECQL4
UniProt ID: (Human) O94761
Entrez Gene ID: (Human) 9401, (Rat) 300057
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.