Invitrogen
Manufacturer Code:PA547495
Catalog # PIPA547495
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse RBP4 Glu19-Leu201 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Plasma retinol-binding protein; PRBP; PRBP RBP plasma retinol-binding protein retinol-binding protein 4 retinol-binding protein 4 interstitial; RBP; retinol binding protein 4, cellular; Retinol-binding protein 4
Gene Aliases: Rbp-4; Rbp4
UniProt ID: (Mouse) Q00724
Entrez Gene ID: (Mouse) 19662
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.