Abnova
Manufacturer Code:PAB15711
Catalog # 89-117-776
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant GST fusion protein corresponding to 117 mouse Rb1cc1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant Rb1cc1.
For Research Use Only
Protein Aliases: 200 kDa FAK family kinase-interacting protein; 2900055E04Rik; 5930404L04Rik; Cc1; coiled coil forming protein 1; Coiled-coil-forming protein 1; DRAGOU14; FAK family kinase-interacting protein of 200 kDa; FIP200; LaXp180; RB1-inducible coiled-coil protein 1
Gene Aliases: 2900055E04Rik; 5930404L04Rik; Cc1; FIP200; Kiaa0203; LaXp180; Rb1cc1
UniProt ID: (Mouse) Q9ESK9
Entrez Gene ID: (Mouse) 12421
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.