Abnova
Manufacturer Code:MAB0005
Catalog # 89-107-816
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5H12 |
Immunogen | Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S807 of human Rb. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against synthetic phosphopeptide of Rb.
For Research Use Only
Protein Aliases: exon 17 tumor GOS561 substitution mutation causes premature stop; GOS563 exon 17 substitution mutation causes premature stop; OSRC; p105-Rb; p110-RB1; pp110; pRb; prepro-retinoblastoma-associated protein; protein phosphatase 1, regulatory subunit 130; RB; retinoblastoma 1; retinoblastoma suspectibility protein; Retinoblastoma-associated protein
Gene Aliases: OSRC; p105-Rb; pp110; PPP1R130; pRb; RB; RB1
UniProt ID: (Human) P06400
Entrez Gene ID: (Human) 5925
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.