Abnova
Manufacturer Code:H00010325D01P
Catalog # 89-018-216
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RRAGB (NP_057740.2 1 a.a. ~ 374 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human RRAGB protein.
For Research Use Only
Protein Aliases: bA465E19.1; GTP-binding protein ragB; Rag B; RAGB; Ras-related GTP binding B; Ras-related GTP-binding protein B; RRAGB
Gene Aliases: bA465E19.1; RAGB; RRAGB
UniProt ID: (Human) Q5VZM2
Entrez Gene ID: (Human) 10325
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.