Invitrogen
Manufacturer Code:MA537663
Catalog # PIMA537663
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1411CT316.2.151.34 |
Immunogen | Raptor recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 4932417H02Rik KIAA1303 KOG1 Mip1 mKIAA1303 p150 target of rapamycin (TOR)-scaffold protein p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats Rap Raptor regulatory associated protein of MTOR complex 1 regulatory associated protein of MTOR complex 1 regulatory-associated protein of mTOR RGD1311784 RPTOR; p150 target of rapamycin (TOR)-scaffold protein; p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats; Raptor; regulatory associated protein of MTOR, complex 1; Regulatory-associated protein of mTOR
Gene Aliases: 4932417H02Rik; KIAA1303; KOG1; Mip1; mKIAA1303; Rap; RAPTOR; RGD1311784; RPTOR
UniProt ID: (Human) Q8N122
Entrez Gene ID: (Human) 57521, (Mouse) 74370, (Rat) 287871
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.