Invitrogen
Manufacturer Code:PA5106370
Catalog # PIPA5106370
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RAP1B corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2810443E11Rik GTP-binding protein smg p21B K-REV OK/SW-cl.11 RAL1B RAP1B RAP1B member of RAS oncogene family Ras family small GTP binding protein RAP1B RAS related protein 1b RAS-related protein RAP1B ras-related protein Rap-1b small GTP binding protein; GTP-binding protein smg p21B; Ras family small GTP binding protein RAP1B; RAS related protein 1b; Ras-related protein Rap-1b; RAS-related protein RAP1B; small GTP binding protein
Gene Aliases: 2810443E11Rik; K-REV; OK/SW-cl.11; RAL1B; RAP1B
UniProt ID: (Human) P61224, (Rat) Q62636, (Mouse) Q99JI6
Entrez Gene ID: (Human) 5908, (Rat) 171337, (Mouse) 215449
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.