Novus Biologicals
Manufacturer Code:NB1202938
Catalog # NB1202938
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues M(1) R R S K A D V E R Y I A S V Q G S(18) of human RanBP2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 358 kDa nucleoporin; acute necrotizing encephalopathy 1 (autosomal dominant); ANE1 E3 SUMO-protein ligase RanBP2 EC 5.2.1.8 Nuclear pore complex protein Nup358 nucleoporin 358 Nucleoporin Nup358 NUP358TRP2 P270 RAN binding protein 2 Ran-binding protein 2 RanBP2 transformation-related protein 2358 kDa nucleoporin TRP1; E3 SUMO-protein ligase RanBP2; Nuclear pore complex protein Nup358; nucleoporin 358; Nucleoporin Nup358; P270; Ran-binding protein 2; RanBP2; transformation-related protein 2
Gene Aliases: ADANE; ANE1; IIAE3; NUP358; RANBP2; TRP1; TRP2
UniProt ID: (Human) P49792
Entrez Gene ID: (Human) 5903
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.