Novus Biologicals
Manufacturer Code:NBP131776
Catalog # NBP131776
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human RAN. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Androgen receptor-associated protein 24; Androgen receptor-associated protein 24 ARA24Gsp1 GTPase Ran GTP-binding nuclear protein Ran guanosine triphosphatase Ran member RAS oncogene family OK/SW-cl.81 RAN member RAS oncogene family RanGTPase Ras-like protein TC4 Ras-related nuclear protein TC4; GTP-binding nuclear protein Ran; GTPase Ran; guanosine triphosphatase Ran; member RAS oncogene family; OK/SW-cl.81; RanGTPase; Ras-like protein TC4; Ras-related nuclear protein
Gene Aliases: ARA24; Gsp1; OK/SW-cl.81; RAN; TC4
UniProt ID: (Human) Q9CZA2
Entrez Gene ID: (Human) 5901
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.