Invitrogen
Manufacturer Code:PA518958
Catalog # PIPA518958
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (DYQSKRTEGIV) corresponding to the C-terminus amino acids of Ramp1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Ramp1 receptor (calcitonin) activity modifying protein 1 9130218E19Rik MGC38240 receptor-activity modifying protein 1; receptor (calcitonin) activity modifying protein 1; Receptor activity-modifying protein 1
Gene Aliases: Ramp1
UniProt ID: (Rat) Q9JJ74
Entrez Gene ID: (Rat) 58965
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.