Novus Biologicals
Manufacturer Code:NBP273785JF549
Catalog # N273785J549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2C4 |
Immunogen | Full length recombinant protein of human RalB (NP_002872) produced in HEK293T cells. |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: GTP binding protein RAS-like protein B ras-related protein Ral-B v-ral simian leukemia viral oncogene homolog B (ras related GTP bindingprotein); ras related GTP binding protein B; RAS-like protein B; Ras-related protein Ral-B; v-ral simian leukemia viral oncogene homolog B (ras related; GTP binding protein)
Gene Aliases: RALB
UniProt ID: (Human) P11234
Entrez Gene ID: (Human) 5899
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.