Invitrogen
Manufacturer Code:MA514886
Catalog # PIMA514886
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | K.353.0 |
Immunogen | Synthetic peptide corresponding to residues near the carboxy terminus of human radixin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: ESP10; Radixin; radixin isoform
Gene Aliases: AA516625; DFNB24; RDX
UniProt ID: (Human) P35241, (Mouse) P26043
Entrez Gene ID: (Human) 5962, (Rat) 315655, (Mouse) 19684
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.