Novus Biologicals
Manufacturer Code:NBP273782APCCY7
Catalog # N273782APC7
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7H4 |
Immunogen | Recombinant protein fragment corresponding to amino acids 119-346 of human Rad51L1 (NP_598193) produced in E.coli. |
Conjugate | APC-Cyanine7 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: DNA repair protein RAD51 homolog 2; DNA repair protein RAD51 homolog 2 MGC34245 R51H2REC2hREC2 RAD51 (S. cerevisiae)-like 1 RAD51 homolog B RAD51B RAD51-like 1 (S. cerevisiae) RAD51-like protein 1 RecA-like protein recombination repair protein; R51H2; RAD51 homolog B; RAD51-like protein 1; Rad51B; RecA-like protein; recombination repair protein
Gene Aliases: R51H2; RAD51B; RAD51L1; REC2
UniProt ID: (Human) O15315
Entrez Gene ID: (Human) 5890
Molecular Function:
DNA binding protein
DNA strand-pairing protein
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.