Novus Biologicals
Manufacturer Code:NB100178
Catalog # NB100178
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5B3/6 |
Immunogen | His-tagged human Rad51D overexpressed in E. coli. [UniProt# O75771] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DNA repair protein RAD51 homolog 4; DNA repair protein RAD51 homolog 4 HsTRAD R51H3Trad RAD51 homolog D RAD51DRAD51 (S. cerevisiae)-like 3 RAD51-like 3 (S. cerevisiae) RAD51-like protein 3 recombination repair protein TRAD; R51H3; RAD51 homolog D; RAD51-like protein 3; recombination repair protein; TRAD
Gene Aliases: BROVCA4; R51H3; RAD51D; RAD51L3; TRAD
UniProt ID: (Human) O75771
Entrez Gene ID: (Human) 5892
Molecular Function:
DNA binding protein
DNA strand-pairing protein
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.