Proteintech
Manufacturer Code:111261AP150UL
Catalog # 50-172-6898
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Rad51D Fusion Protein Ag1595 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA repair protein RAD51 homolog 4; HsTRAD R51H3 RAD51 homolog D RAD51 like 3 (S. cerevisiae) RAD51 like protein 3 Rad51D RAD51L3 Trad; R51H3; RAD51 homolog D; RAD51-like 3; RAD51-like protein 3; recombination repair protein; TRAD
Gene Aliases: BROVCA4; R51H3; RAD51D; RAD51L3; TRAD
UniProt ID: (Human) O75771, (Mouse) O55230
Entrez Gene ID: (Human) 5892, (Rat) 303375, (Mouse) 19364
Molecular Function:
DNA binding protein
DNA strand-pairing protein
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.