Novus Biologicals
Manufacturer Code:NB100177
Catalog # NB100177
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2H11/6 |
Immunogen | His-tagged human Rad51C over-expressed in E. coli. [UniProt# O43502] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BROVCA3 DNA repair protein RAD51 homolog 3 FANCO MGC104277 R51H3 RAD51 homolog C RAD51 homolog C (S. cerevisiae) RAD51L2RAD51 (S. cerevisiae) homolog C RAD51-like protein 2 yeast RAD51 homolog 3; DNA repair protein RAD51 homolog 3; R51H3; RAD51 homolog C; RAD51-like protein 2; yeast RAD51 homolog 3
Gene Aliases: BROVCA3; FANCO; R51H3; RAD51C; RAD51L2
UniProt ID: (Human) O43502
Entrez Gene ID: (Human) 5889
Molecular Function:
DNA binding protein
DNA strand-pairing protein
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.