Novus Biologicals
Manufacturer Code:NBP275640
Catalog # NBP275640
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JM54-26 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: BRCA1/BRCA2-containing complex, subunit 5; BRCC5 HRAD51 HsRAD51 HsT16930 RAD51 (S. cerevisiae) homolog (E coli RecA homolog) RAD51 homolog (RecA homolog E. coli) (S. cerevisiae) RAD51 homolog A RAD51ABRCA1/BRCA2-containing complex subunit 5 RecA E. coli homolog of RECADNA repair protein RAD51 homolog 1 RecA-like protein recombination protein A; DNA repair protein RAD51 homolog 1; HsRAD51; RAD51 homolog A; RecA, E. coli, homolog of; RecA-like protein; recombination protein A
Gene Aliases: BRCC5; FANCR; HRAD51; HsRad51; HsT16930; MRMV2; RAD51; RAD51A; RECA
UniProt ID: (Human) Q06609
Entrez Gene ID: (Human) 5888
Molecular Function:
DNA binding protein
DNA strand-pairing protein
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.