R&D Systems
Manufacturer Code:MAB4996
Catalog # MAB4996
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 684945 |
Immunogen | <em>E. coli-</em>derived recombinant human Rad50 Leu518-Leu881 Accession # Q92878 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DNA repair protein RAD50; DNA repair protein RAD50 EC 3.6 EC 3.6.1.15 EC 3.6.3 EC 3.6.3.27 hRAD50 NBSLD RAD50 (S. cerevisiae) homolog RAD50 homolog (S. cerevisiae) RAD502 RAD50-2; hRAD50; RAD50 homolog, double strand break repair protein
Gene Aliases: hRad50; NBSLD; RAD50; RAD502
UniProt ID: (Human) Q92878
Entrez Gene ID: (Human) 10111
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.