Novus Biologicals
Manufacturer Code:NB100154
Catalog # NB100154
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C-terminal mouse RAD50 (604 amino acids). [UniProt# P70388] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA repair protein RAD50; DNA repair protein RAD50 EC 3.6 EC 3.6.1.15 EC 3.6.3 EC 3.6.3.27 hRAD50 NBSLD RAD50 (S. cerevisiae) homolog RAD50 homolog (S. cerevisiae) RAD502 RAD50-2; hRAD50; RAD50 homolog, double strand break repair protein
Gene Aliases: hRad50; NBSLD; RAD50; RAD502
UniProt ID: (Human) Q92878
Entrez Gene ID: (Human) 10111
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.