Novus Biologicals
Manufacturer Code:NB100386
Catalog # NB100386
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CM110-2C10 |
Immunogen | An N-terminal fusion protein of the C-terminal fragment of Scc1 subunit human cohesin (amino acids 544-631 of hRad21). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 64-kDa C-terminal product; 64-kDa carboxy-terminal product; 65-kDa carboxy-terminal product; Double-strand-break repair protein rad21 homolog; double-strand-break repair protein rad21 homolog hHR21FLJ25655 HR21 KIAA0078RAD21 (S. pombe) homolog MCD1 Nuclear matrix protein 1 NXP-1 NXP1HRAD21 protein involved in DNA double-strand break repair RAD21 homolog (S. pombe) SCC1 homolog SCC1FLJ40596; hHR21; kleisin; Nuclear matrix protein 1; NXP-1; protein involved in DNA double-strand break repair; RAD21 homolog; SCC1 homolog; sister chromatid cohesion 1
Gene Aliases: CDLS4; hHR21; HR21; HRAD21; KIAA0078; MCD1; NXP1; RAD21; SCC1
UniProt ID: (Human) O60216
Entrez Gene ID: (Human) 5885
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.