Novus Biologicals
Manufacturer Code:NBP293566100UL
Catalog # NB029005
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-282 of human RAD1 (NP_002844.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cell cycle checkpoint protein Hrad1; cell cycle checkpoint protein Hrad1 checkpoint control protein HRAD1 DNA repair exonuclease rad1 homolog DNA repair exonuclease REC1 EC 3.1.11.2 exonuclease homolog RAD1 HRAD1 RAD1 (S. pombe) homolog RAD1 homolog (S. pombe) Rad1-like DNA damage checkpoint protein REC1cell cycle checkpoint protein RAD1; Cell cycle checkpoint protein RAD1; checkpoint control protein HRAD1; DNA repair exonuclease rad1 homolog; DNA repair exonuclease REC1; exonuclease homolog RAD1; hRAD1; RAD1 checkpoint clamp component; RAD1 homolog; Rad1-like DNA damage checkpoint protein
Gene Aliases: HRAD1; RAD1; REC1
UniProt ID: (Human) O60671
Entrez Gene ID: (Human) 5810
Molecular Function:
DNA binding protein
exodeoxyribonuclease
hydrolase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.