Novus Biologicals
Manufacturer Code:NBP267091
Catalog # NBP267091
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JM11-29 |
Immunogen | Recombinant full length protein of human RAC1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: Cell migration-inducing gene 5 protein; Cell migration-inducing gene 5 protein MGC111543 p21-Rac1migration-inducing gene 5 Ras-like protein TC25 ras-related C3 botulinum toxin substrate 1 ras-related C3 botulinum toxin substrate 1 (rho family small GTP bindingprotein Rac1) rho family small GTP binding protein Rac1 TC25 TC-25; p21-Rac1; Ras-like protein TC25; Ras-related C3 botulinum toxin substrate 1
Gene Aliases: MIG5; p21-Rac1; Rac-1; RAC1; TC-25; TC25
UniProt ID: (Human) P63000
Entrez Gene ID: (Human) 5879
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.