Proteintech
Manufacturer Code:240721AP150UL
Catalog # 50-173-4231
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Rac1 Fusion Protein Ag17779 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell migration-inducing gene 5 protein; MIG5 p21 Rac1 RAC1 RAC1b Ras like protein TC25 TC 25 TC25; p21-Rac1; Ras-like protein TC25; Ras-related C3 botulinum toxin substrate 1; ras-related C3 botulinum toxin substrate 1 (rho family small GTP binding protein Rac1); rho family, small GTP binding protein Rac1
Gene Aliases: AL023026; D5Ertd559e; MIG5; p21-Rac1; Rac-1; RAC1; TC-25; TC25
UniProt ID: (Human) P63000, (Mouse) P63001, (Rat) Q6RUV5
Entrez Gene ID: (Human) 5879, (Mouse) 19353, (Rat) 363875
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.