Novus Biologicals
Manufacturer Code:NBP169184
Catalog # NBP169184
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to Rph3a (rabphilin 3A) The peptide sequence was selected from the N terminal of Rph3a. Peptide sequence GAWFFKGFPKQVLPQPMPIKKTKPQQPAGEPATQEQPTPESRHPARAPAR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Exophilin-1; Exophilin-1 KIAA0985exophilin-1 rabphilin rabphilin 3A homolog (mouse) rabphilin-3A; rabphilin 3A homolog; Rabphilin-3A
Gene Aliases: KIAA0985; RPH3A
UniProt ID: (Human) Q9Y2J0
Entrez Gene ID: (Human) 22895
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.