EMD Millipore
Manufacturer Code:041657
Catalog # 04-165-7MI
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Secondary Antibody |
Clone | RM125 |
Immunogen | Human IgA2 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Stable for one year at 2°C to 8°C from date of receipt. Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage. |
Target | IgG |
Rabbit Monoclonal Antibody
For Research Use Only
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.