Invitrogen
Manufacturer Code:A27019
Catalog # PIA27019
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Goat |
Host / Isotype | Rabbit |
Class | Oligoclonal |
Type | Secondary Antibody |
Immunogen | Recombit full-length protein |
Conjugate | Alexa Fluor 790 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Target | IgG |
Rabbit Recombinant Polyclonal Secondary Antibody
For Research Use Only
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.