Novus Biologicals
Manufacturer Code:NBP298732AF594
Catalog # NB105359
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Rab7a. |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CMT2B FLJ20819 member RAS oncogene family PSN RAB7A member RAS oncogene family Ras-associated protein RAB7 ras-related protein Rab-7a; RAB7, member RAS oncogene family; Ras-associated protein RAB7; Ras-related protein Rab-7a
Gene Aliases: PRO2706; RAB7; RAB7A
UniProt ID: (Human) P51149
Entrez Gene ID: (Human) 7879
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.