Invitrogen
Manufacturer Code:MA517161
Catalog # PIMA517161
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4E11 |
Immunogen | Purified recombit fragment of human RAB4A expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HTLV-1 related endogenous sequence; Ras-related protein Rab-4A; Ras-related small GTP binding protein 4
Gene Aliases: AI848268; HRES-1; HRES-1/RAB4; HRES1; RAB4; RAB4A
UniProt ID: (Human) P20338, (Rat) P05714, (Mouse) P56371
Entrez Gene ID: (Human) 5867, (Rat) 25532, (Mouse) 19341
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.