Novus Biologicals
Manufacturer Code:NBP273773IR
Catalog # NBP273773IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1G3 |
Immunogen | Full length recombinant protein of human Rab3A (NP_002857) produced in E.coli. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: RAB3A member RAS oncogene family RAS-associated protein RAB3A ras-related protein Rab-3A; RAS-associated protein RAB3A; Ras-related protein Rab-3A
Gene Aliases: RAB3A
UniProt ID: (Human) P20336
Entrez Gene ID: (Human) 5864
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.