Novus Biologicals
Manufacturer Code:NBP237241
Catalog # NBP237241
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4D3F11 |
Immunogen | Purified recombinant fragment of human RAB27A (AA: FULL(1-221)) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: GS2rab-27 GTP-binding protein Ram HsT18676 Rab-27 RAB27A member RAS oncogene family RAB27MGC117246 RAMras-related protein Rab-27A; GTP-binding protein Ram; mutant Ras-related protein Rab-27A; Rab-27; Ras-related protein Rab-27A
Gene Aliases: GS2; HsT18676; RAB27; RAB27A; RAM
UniProt ID: (Human) P51159
Entrez Gene ID: (Human) 5873
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.