R&D Systems
Manufacturer Code:AF7245SP
Catalog # AF7245SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human RAB27A Ser135-Ala218 Accession # P51159 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: GS2 GS2rab-27 GTP-binding protein Ram HsT18676 Rab-27 RAB27A member RAS oncogene family RAB27MGC117246 RAM RAMras-related protein Rab-27A; GTP-binding protein Ram; mutant Ras-related protein Rab-27A; Rab-27; Ras-related protein Rab-27A
Gene Aliases: GS2; HsT18676; RAB27; RAB27A; RAM
UniProt ID: (Human) P51159
Entrez Gene ID: (Human) 5873
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.