Novus Biologicals
Manufacturer Code:NBP299499R
Catalog # NB085417
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Rab1A. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp564B163 member RAS oncogene family RAB1A member RAS oncogene family ras-related protein Rab-1A; GTP binding protein Rab1a; Rab GTPase YPT1 homolog; RAB1, member RAS oncogene family; Ras-related protein Rab-1A; YPT1-related protein
Gene Aliases: RAB1; RAB1A; YPT1
UniProt ID: (Human) P62820
Entrez Gene ID: (Human) 5861
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.