Novus Biologicals
Manufacturer Code:NBP29320120UL
Catalog # NB028290
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-203 of human RAB13 (NP_002861.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell growth-inhibiting gene 4 protein; Cell growth-inhibiting gene 4 protein growth-inhibiting gene 4 protein RAB13 member RAS oncogene family RAS-associated protein RAB13 ras-related protein Rab-13; growth-inhibiting gene 4 protein; RAS-associated protein RAB13; Ras-related protein Rab-13
Gene Aliases: GIG4; RAB13
UniProt ID: (Human) Q5U0A6
Entrez Gene ID: (Human) 5872
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.