Novus Biologicals
Manufacturer Code:NB1001086
Catalog # NB1001086
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence TENKPKVQCCQNI corresponding to C-Terminus according to NP_004654.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC1490 RAB 11A member oncogene family Rab-11 RAB11 RAB11A member RAS oncogene family YL8ras-related protein Rab-11A; RAB 11A, member oncogene family; Rab-11; Ras-related protein Rab-11A; YL8
Gene Aliases: RAB11; RAB11A; YL8
UniProt ID: (Human) P62491
Entrez Gene ID: (Human) 8766
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.