Abnova
Manufacturer Code:PAB17291
Catalog # 89-118-955
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal of human RYR2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of RYR2.
For Research Use Only
Protein Aliases: ARVC2; ARVD2; Cardiac muscle ryanodine receptor; Cardiac muscle ryanodine receptor-calcium release channel; cardiac-type ryanodine receptor; islet-type ryanodine receptor; kidney-type ryanodine receptor; Ryanodine receptor 2; ryanodine receptor 2 (cardiac); RYR-2; Type 2 ryanodine receptor; VTSIP
Gene Aliases: ARVC2; ARVD2; RyR; RYR-2; RYR2; VTSIP
UniProt ID: (Human) Q92736
Entrez Gene ID: (Human) 6262
Molecular Function:
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.