Proteintech
Manufacturer Code:665391IG150UL
Catalog # 50-173-6959
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1B4E12 |
Immunogen | RYR1 Fusion Protein Ag25120 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: calcium release channel; CCO MHS MHS1 Ryanodine receptor 1 RYDR RYR RYR 1 RYR1 SKRR; central core disease of muscle; protein phosphatase 1, regulatory subunit 137; Ryanodine receptor 1; ryanodine receptor 1 (skeletal); RYR-1; sarcoplasmic reticulum calcium release channel; Skeletal muscle calcium release channel; Skeletal muscle ryanodine receptor; Skeletal muscle-type ryanodine receptor; Type 1 ryanodine receptor; type 1-like ryanodine receptor
Gene Aliases: AI528790; CCO; MHS; MHS1; PPP1R137; RYDR; RYR; RYR-1; RYR1; SKRR
UniProt ID: (Human) P21817, (Mouse) E9PZQ0
Entrez Gene ID: (Human) 6261, (Mouse) 20190
Molecular Function: ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.