Abnova
Manufacturer Code:PAB5562
Catalog # 89-131-909
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of human RYR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of RYR1.
For Research Use Only
Protein Aliases: CCO; central core disease of muscle; MHS; MHS1; protein phosphatase 1, regulatory subunit 137; Ryanodine receptor 1; ryanodine receptor 1 (skeletal); RYDR; RYR; RYR-1; sarcoplasmic reticulum calcium release channel; Skeletal muscle calcium release channel; Skeletal muscle ryanodine receptor; Skeletal muscle-type ryanodine receptor; SKRR; Type 1 ryanodine receptor; type 1-like ryanodine receptor
Gene Aliases: CCO; MHS; MHS1; PPP1R137; RYDR; RYR; RYR-1; RYR1; SKRR
UniProt ID: (Human) P21817
Entrez Gene ID: (Human) 6261
Molecular Function: ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.