Invitrogen
Manufacturer Code:PA514734
Catalog # PIPA514734
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 561-591 amino acids from the C-terminal region of human RYK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hydroxyaryl-protein kinase; JTK5A protein tyrosine kinase; Tyrosine-protein kinase RYK; Tyrosine-protein kinase RYK precursor
Gene Aliases: D3S3195; JTK5; JTK5A; RYK; RYK1
UniProt ID: (Human) P34925
Entrez Gene ID: (Human) 6259
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.