Invitrogen
Manufacturer Code:PA514733
Catalog # PIPA514733
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 160-190 amino acids from human RYK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: growth factor receptor; hydroxyaryl-protein kinase; JTK5A protein tyrosine kinase; Kinase VIK; Met-related kinase; NYK-R; Tyrosine-protein kinase RYK; Tyrosine-protein kinase RYK precursor
Gene Aliases: AW536699; D3S3195; ERK-3; JTK5; JTK5A; Mrk; RYK; RYK1; Vik
UniProt ID: (Human) P34925, (Mouse) Q01887
Entrez Gene ID: (Human) 6259, (Rat) 140585, (Mouse) 20187
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.