Invitrogen
Manufacturer Code:PA130319
Catalog # PIPA130319
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH corresponding to amino acids 175-191 of Human RYK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: growth factor receptor; hydroxyaryl-protein kinase; JTK5A protein tyrosine kinase; Kinase VIK; Met-related kinase; NYK-R; RYK1 Receptor-like tyrosine kinase D3S3195 ERK3 Hydroxyaryl-protein kinase; Tyrosine-protein kinase RYK
Gene Aliases: AW536699; D3S3195; ERK-3; JTK5; JTK5A; Mrk; RYK; RYK1; Vik
UniProt ID: (Human) P34925, (Mouse) Q01887
Entrez Gene ID: (Human) 6259, (Mouse) 20187
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.