Abnova
Manufacturer Code:H00006259A01
Catalog # 89-105-383
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 54 a.a. &bsim 163 a.a) partial recombinant protein with GST tag.,RYK (NP_002949 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant RYK.
For Research Use Only
Protein Aliases: D3S3195; hydroxyaryl-protein kinase; JTK5; JTK5A; JTK5A protein tyrosine kinase; RYK1; Tyrosine-protein kinase RYK
Gene Aliases: D3S3195; JTK5; JTK5A; RYK; RYK1
UniProt ID: (Human) P34925
Entrez Gene ID: (Human) 6259
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.