Invitrogen
Manufacturer Code:PA518311
Catalog # PIPA518311
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (RQRSRERAESEAEC) corresponding to the internal amino acids of RXRG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Nuclear receptor subfamily 2 group B member 3; Retinoic acid receptor RXR-gamma; Retinoid X receptor gamma; retinoid X receptor, gamma; RXRG RXRC NR2B3 retinoid X receptor gamma
Gene Aliases: NR2B3; RXRC; RXRG
UniProt ID: (Human) P48443
Entrez Gene ID: (Human) 6258
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.