Abnova
Manufacturer Code:PAB6694
Catalog # 89-132-607
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human RXRG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of RXRG.
For Research Use Only
Protein Aliases: NR2B3; Nuclear receptor subfamily 2 group B member 3; Retinoic acid receptor RXR-gamma; Retinoid X receptor gamma; retinoid X receptor, gamma; RXRC
Gene Aliases: NR2B3; RXRC; RXRG
UniProt ID: (Human) P48443
Entrez Gene ID: (Human) 6258
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.